Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266120.1 | NP_001253049.1 | 851 | 88 | ||
| GenBank mRNA | JU335476.1 | - | 880 | 88 | ||
| JU472348.1 | - | 972 | 88 | |||
| JV045586.1 | - | 851 | 88 | |||
More Information
Set Membership: |
|

UBFD1
Genomic Map