Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266305.1 | NP_001253234.1 | 161 | 91 | ||
| GenBank mRNA | JU335038.1 | - | 161 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PMP2| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266305.1 | NP_001253234.1 | 161 | 91 | ||
| GenBank mRNA | JU335038.1 | - | 161 | 91 | ||