Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266763.2 | NP_001253692.1 | 365 | 108 | ||
| XM_015146202.2 | XP_015001688.1 | 352 | 108 | |||
| GenBank mRNA | JV599337.1 | - | 3961 | 108 | ||
| JV600284.1 | - | 365 | 108 | |||
| Gene Symbol: C8H8orf82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV617303.1 | - | 1386 | 108 | ||
More Information
Set Membership: |
|

LRRC24
Genomic Map