Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266825.1 | NP_001253754.1 | 1169 | 69 | ||
| XM_015141097.2 | XP_014996583.1 | 1481 | 69 | |||
| XM_015141098.2 | XP_014996584.1 | 2023 | 69 | |||
| XM_015141099.2 | XP_014996585.1 | 1438 | 69 | |||
| GenBank mRNA | JU321968.1 | - | 1169 | 69 | ||
More Information
Set Membership: |
|

KCTD16
Genomic Map