Assay Details
| Gene Symbol: DCAF8L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266964.1 | NP_001253893.1 | 224 | 86 | ||
| XM_015127118.2 | XP_014982604.1 | 616 | 86 | |||
| XM_028841748.1 | XP_028697581.1 | 693 | 86 | |||
| XM_028841749.1 | XP_028697582.1 | 800 | 86 | |||
| XM_028841750.1 | XP_028697583.1 | 533 | 86 | |||
| GenBank mRNA | JV046959.1 | - | 224 | 86 | ||
More Information
Set Membership: |
|

DCAF8L2
Genomic Map