Assay Details
| Gene Symbol: FCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261542.1 | NP_001248471.1 | 1018 | 115 | ||
| GenBank mRNA | JU334772.1 | - | 1039 | 115 | ||
| JU472388.1 | - | 1034 | 115 | |||
| JV047237.1 | - | 1022 | 115 | |||
| JV047239.1 | - | 1018 | 115 | |||
| JV716246.1 | - | 1040 | 115 | |||
More Information
Set Membership: |
|

FCF1
Genomic Map