Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261620.1 | NP_001248549.1 | 1324 | 104 | ||
| XM_015128220.2 | XP_014983706.1 | 1259 | 104 | |||
| GenBank mRNA | JU328250.1 | - | 844 | 104 | ||
| JV047663.1 | - | 1324 | 104 | |||
More Information
Set Membership: |
|

MOSPD1
Genomic Map