Assay Details
| Gene Symbol: RPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266200.1 | NP_001253129.1 | 1325 | 97 | ||
| GenBank mRNA | JV565044.1 | - | 1325 | 97 | ||
| JV658103.1 | - | 1331 | 97 | |||
More Information
Set Membership: |
|

RPL4
Genomic Map