Assay Details
| Gene Symbol: POLR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257459.1 | NP_001244388.1 | 615 | 89 | ||
| GenBank mRNA | JU321909.1 | - | 615 | 89 | ||
| JU473090.1 | - | 595 | 89 | |||
| JV044812.1 | - | 622 | 89 | |||
More Information
Set Membership: |
|

POLR1D
Genomic Map