Assay Details
| Gene Symbol: CALHM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111367.4 | XP_001111367.2 | 560 | 94 | ||
| XM_028847054.1 | XP_028702887.1 | 184 | 94 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALHM4| Gene Symbol: CALHM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111367.4 | XP_001111367.2 | 560 | 94 | ||
| XM_028847054.1 | XP_028702887.1 | 184 | 94 | |||
Set Membership: |
|