Assay Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798535.3 | XP_002798581.1 | 1938 | 71 | ||
| XM_015151488.2 | XP_015006974.1 | 1854 | 71 | |||
| XM_015151489.2 | XP_015006975.1 | 1988 | 71 | |||
| XM_028829410.1 | XP_028685243.1 | 1935 | 71 | |||
| GenBank mRNA | JU322132.1 | - | 1849 | 71 | ||
| JV046245.1 | - | 1738 | 71 | |||
More Information
Set Membership: |
|

PRICKLE1
Genomic Map