Assay Details
| Gene Symbol: ASCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094978.3 | XP_001094978.1 | 1379 | 66 | ||
| GenBank mRNA | JU327049.1 | - | 589 | 66 | ||
| JV715316.1 | - | 555 | 66 | |||
More Information
Set Membership: |
|

ASCL1
Genomic Map