Assay Details
| Gene Symbol: LOC717879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136790.2 | XP_014992276.1 | 362 | 104 | ||
| GenBank mRNA | JU474344.1 | - | 240 | 104 | ||
| JV704290.1 | - | 262 | 104 | |||
| Gene Symbol: LOC114674848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842063.1 | XP_028697896.1 | 151 | 104 | ||
More Information
Set Membership: |
|

LOC717879
Genomic Map