Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111887.4 | XP_001111887.2 | 894 | 78 | ||
| XM_028834141.1 | XP_028689974.1 | 1564 | 78 | |||
| GenBank mRNA | JU336604.1 | - | 443 | 78 | ||
| JU477419.1 | - | 453 | 78 | |||
| JV048385.1 | - | 441 | 78 | |||
More Information
Set Membership: |
|

PUS3
Genomic Map