Assay Details
| Gene Symbol: SLC52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090939.4 | XP_001090939.2 | 879 | 70 | ||
| XM_015146279.2 | XP_015001765.2 | 422 | 70 | |||
| XM_015146280.2 | XP_015001766.2 | 581 | 70 | |||
| XM_028853123.1 | XP_028708956.1 | 875 | 70 | |||
| GenBank mRNA | JU336378.1 | - | 238 | 70 | ||
| JU477196.1 | - | 245 | 70 | |||
| JV048247.1 | - | 184 | 70 | |||
| JV661359.1 | - | 245 | 70 | |||
More Information
Set Membership: |
|

SLC52A2
Genomic Map