Assay Details
| Gene Symbol: ZNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125448.2 | XP_014980934.2 | 2291 | 76 | ||
| XM_028841457.1 | XP_028697290.1 | 1273 | 76 | |||
| GenBank mRNA | JU332182.1 | - | 1097 | 76 | ||
| JV639191.1 | - | 1157 | 76 | |||
More Information
Set Membership: |
|

ZNF213
Genomic Map