Assay Details
| Gene Symbol: FLVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144152.2 | XP_014999638.1 | 1908 | 63 | ||
| XM_015144154.2 | XP_014999640.1 | 1199 | 63 | |||
| XM_028851701.1 | XP_028707534.1 | 1767 | 63 | |||
| XM_028851702.1 | XP_028707535.1 | 1476 | 63 | |||
More Information
Set Membership: |
|

FLVCR2
Genomic Map