Assay Details
| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117747.4 | XP_001117747.2 | 1419 | 66 | ||
| GenBank mRNA | JU346600.1 | - | 734 | 66 | ||
| JU476598.1 | - | 903 | 66 | |||
More Information
Set Membership: |
|

CXCL16
Genomic Map