Assay Details
| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112062.4 | XP_001112062.1 | 503 | 75 | ||
| XM_028833760.1 | XP_028689593.1 | 358 | 75 | |||
More Information
Set Membership: |
|

SHISAL2A
Genomic Map