Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137680.2 | XP_014993166.1 | 3085 | 63 | ||
| XM_015137687.2 | XP_014993173.1 | 1191 | 63 | |||
| XM_028834627.1 | XP_028690460.1 | 939 | 63 | |||
| GenBank mRNA | JU323038.1 | - | 1510 | 63 | ||
| JV708621.1 | - | 1505 | 63 | |||
More Information
Set Membership: |
|

DPH2
Genomic Map