Assay Details
| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116165.2 | XP_014971651.1 | 280 | 76 | ||
| XM_028832935.1 | XP_028688768.1 | 819 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PATE1| Gene Symbol: PATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116165.2 | XP_014971651.1 | 280 | 76 | ||
| XM_028832935.1 | XP_028688768.1 | 819 | 76 | |||
Set Membership: |
|