Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118415.2 | XP_014973901.2 | 1277 | 86 | ||
| GenBank mRNA | JV053306.1 | - | 266 | 86 | ||
| JV643812.1 | - | 329 | 86 | |||
More Information
Set Membership: |
|

SLC16A11
Genomic Map