Assay Details
| Gene Symbol: PRRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092489.4 | XP_001092489.3 | 435 | 85 | ||
| XM_015128426.2 | XP_014983912.1 | 278 | 85 | |||
| XM_028841872.1 | XP_028697705.1 | 262 | 85 | |||
| GenBank mRNA | JV043789.1 | - | 205 | 85 | ||
| JV481514.1 | - | 226 | 85 | |||
More Information
Set Membership: |
|

PRRG3
Genomic Map