Assay Details
| Gene Symbol: MIR514B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128311.1 | 44 | 83 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | 
|---|
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Macaca mulatta | 
 Back To Top
 Back To Top
 MIR514B
MIR514B Genomic Map
Genomic Map Back To Top
 Back To Top