Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194908.1 | NP_001181837.1 | 467 | 120 | ||
| XM_002806144.2 | XP_002806190.1 | 276 | 120 | |||
| XM_028841832.1 | XP_028697665.1 | 791 | 120 | |||
| GenBank mRNA | JU337150.1 | - | 155 | 120 | ||
| JU471908.1 | - | 75 | 120 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194908.1, XM_002806144.2 transcripts.
Set Membership: |
|

RAB9A
Genomic Map