Assay Details
| Gene Symbol: GPR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261006.1 | NP_001247935.1 | 1317 | 96 | ||
| GenBank mRNA | JU320551.1 | - | 1317 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261006.1 transcripts.
Set Membership: |
|

GPR22
Genomic Map