Assay Details
| Gene Symbol: SERTAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261684.1 | NP_001248613.1 | 242 | 92 | ||
| GenBank mRNA | JU322923.1 | - | 159 | 92 | ||
| JU472467.1 | - | 283 | 92 | |||
| JV043908.1 | - | 242 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261684.1 transcripts.
Set Membership: |
|

SERTAD2
Genomic Map