Assay Details
| Gene Symbol: BOLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265651.1 | NP_001252580.1 | 67 | 69 | ||
| XM_015112536.2 | XP_014968022.1 | 188 | 69 | |||
| GenBank mRNA | JU473800.1 | - | 88 | 69 | ||
| JV565023.1 | - | 85 | 69 | |||
| JV667298.1 | - | 67 | 69 | |||
More Information
Set Membership: |
|

BOLA3
Genomic Map