Assay Details
| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266623.1 | NP_001253552.1 | 1263 | 68 | ||
| XM_028841649.1 | XP_028697482.1 | 2141 | 68 | |||
| XM_028841650.1 | XP_028697483.1 | 1461 | 68 | |||
| GenBank mRNA | JU477387.1 | - | 1230 | 68 | ||
| JV048367.1 | - | 1263 | 68 | |||
| JV697868.1 | - | 1306 | 68 | |||
More Information
Set Membership: |
|

AKAP17A
Genomic Map