Assay Details
| Gene Symbol: ZBED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266708.1 | NP_001253637.1 | 134 | 105 | ||
| XM_028841621.1 | XP_028697454.1 | 162 | 105 | |||
| GenBank mRNA | JU471622.1 | - | 135 | 105 | ||
| JU471623.1 | - | 135 | 105 | |||
| JU471624.1 | - | 135 | 105 | |||
| JU471625.1 | - | 135 | 105 | |||
| JU471626.1 | - | 135 | 105 | |||
| JV048673.1 | - | 134 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266708.1, XM_028841621.1 transcripts.
Set Membership: |
|

ZBED1
Genomic Map