Assay Details
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302780.1 | NP_001289709.1 | 822 | 81 | ||
| NM_001302781.1 | NP_001289710.1 | 215 | 81 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMIGD3| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302780.1 | NP_001289709.1 | 822 | 81 | ||
| NM_001302781.1 | NP_001289710.1 | 215 | 81 | |||
Set Membership: |
|