Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265931.1 | NP_001252860.1 | 1720 | 89 | ||
| GenBank mRNA | JU323332.1 | - | 1615 | 89 | ||
| JU477468.1 | - | 1674 | 89 | |||
| JV043833.1 | - | 1720 | 89 | |||
| JV663214.1 | - | 1688 | 89 | |||
More Information
Set Membership: |
|

RNASEH2C
Genomic Map