Assay Details
| Gene Symbol: LOC100429241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318352.1 | NP_001305281.1 | 222 | 93 | ||
| GenBank mRNA | EU934785.1 | - | 222 | 93 | ||
| JQ239036.1 | - | 102 | 93 | |||
| JQ239038.1 | - | 102 | 93 | |||
| KF528937.1 | - | 126 | 93 | |||
| KF880612.1 | - | 213 | 93 | |||
More Information
Set Membership: |
|

LOC100429241
Genomic Map