Assay Details
| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125975.2 | XP_014981461.2 | 798 | 62 | ||
| XM_028841416.1 | XP_028697249.1 | 800 | 62 | |||
| XM_028841418.1 | XP_028697251.1 | 874 | 62 | |||
| XM_028841419.1 | XP_028697252.1 | 791 | 62 | |||
| XM_028841420.1 | XP_028697253.1 | 889 | 62 | |||
| XM_028841421.1 | XP_028697254.1 | 971 | 62 | |||
| XM_028841422.1 | XP_028697255.1 | 924 | 62 | |||
| XM_028841423.1 | XP_028697256.1 | 810 | 62 | |||
| GenBank mRNA | JU321921.1 | - | 728 | 62 | ||
| JU476003.1 | - | 684 | 62 | |||
| JV653731.1 | - | 731 | 62 | |||
More Information
Set Membership: |
|

PHKG2
Genomic Map