Assay Details
| Gene Symbol: PCDHB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141075.2 | XP_014996561.2 | 3849 | 80 | ||
| GenBank mRNA | JU330582.1 | - | 214 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB12| Gene Symbol: PCDHB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141075.2 | XP_014996561.2 | 3849 | 80 | ||
| GenBank mRNA | JU330582.1 | - | 214 | 80 | ||
Set Membership: |
|