Assay Details
| Gene Symbol: OTUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097857.4 | XP_001097857.2 | 1684 | 88 | ||
| GenBank mRNA | JV645220.1 | - | 1018 | 88 | ||
| JV709492.1 | - | 933 | 88 | |||
More Information
Set Membership: |
|

OTUD1
Genomic Map