Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111750.2 | XP_014967236.1 | 422 | 101 | ||
| GenBank mRNA | JU319683.1 | - | 297 | 101 | ||
| JU319684.1 | - | 297 | 101 | |||
| JU319685.1 | - | 297 | 101 | |||
| JU319687.1 | - | 297 | 101 | |||
| JU319688.1 | - | 297 | 101 | |||
| JU332113.1 | - | 297 | 101 | |||
| JV602126.1 | - | 379 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015111750.2 transcripts.
Set Membership: |
|

VSNL1
Genomic Map