Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111750.2 | XP_014967236.1 | 590 | 84 | ||
| XM_015111751.2 | XP_014967237.1 | 611 | 84 | |||
| XM_015111752.2 | XP_014967238.1 | 586 | 84 | |||
| XM_015111753.2 | XP_014967239.1 | 697 | 84 | |||
| XM_028831957.1 | XP_028687790.1 | 1136 | 84 | |||
| XM_028831959.1 | XP_028687792.1 | 186 | 84 | |||
| GenBank mRNA | JU319683.1 | - | 465 | 84 | ||
| JU319684.1 | - | 465 | 84 | |||
| JU319685.1 | - | 465 | 84 | |||
| JU319686.1 | - | 461 | 84 | |||
| JU319687.1 | - | 465 | 84 | |||
| JU319688.1 | - | 465 | 84 | |||
| JU332113.1 | - | 465 | 84 | |||
| JV602126.1 | - | 547 | 84 | |||
More Information
Set Membership: |
|

VSNL1
Genomic Map