Assay Details
| Gene Symbol: TCL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101160.3 | XP_001101160.2 | 679 | 86 | ||
| GenBank mRNA | JV076286.1 | - | 121 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCL1B| Gene Symbol: TCL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101160.3 | XP_001101160.2 | 679 | 86 | ||
| GenBank mRNA | JV076286.1 | - | 121 | 86 | ||
Set Membership: |
|