Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134564.2 | XP_014990050.1 | 1209 | 100 | ||
| XM_015134565.2 | XP_014990051.2 | 1219 | 100 | |||
| GenBank mRNA | JV639929.1 | - | 1058 | 100 | ||
More Information
Set Membership: |
|

LRRC17
Genomic Map