Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132586.2 | XP_014988072.2 | 789 | 84 | ||
| GenBank mRNA | JU474516.1 | - | 1364 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF168| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132586.2 | XP_014988072.2 | 789 | 84 | ||
| GenBank mRNA | JU474516.1 | - | 1364 | 84 | ||
Set Membership: |
|