Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847744.1 | XP_028703577.1 | 1262 | 61 | ||
| GenBank mRNA | JU328787.1 | - | 525 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFP57| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847744.1 | XP_028703577.1 | 1262 | 61 | ||
| GenBank mRNA | JU328787.1 | - | 525 | 61 | ||
Set Membership: |
|