Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140635.2 | XP_014996121.1 | 1046 | 68 | ||
| XM_015140636.2 | XP_014996122.1 | 1144 | 68 | |||
| GenBank mRNA | JU326330.1 | - | 621 | 68 | ||
More Information
Set Membership: |
|

PRR16
Genomic Map