Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827944.1 | XP_028683777.1 | 6273 | 85 | ||
| GenBank mRNA | JV646088.1 | - | 5524 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIC2| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827944.1 | XP_028683777.1 | 6273 | 85 | ||
| GenBank mRNA | JV646088.1 | - | 5524 | 85 | ||
Set Membership: |
|