Assay Details
| Gene Symbol: LRATD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU339470.1 | - | 119 | 150 | ||
| JU339471.1 | - | 119 | 150 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRATD2| Gene Symbol: LRATD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU339470.1 | - | 119 | 150 | ||
| JU339471.1 | - | 119 | 150 | |||
Set Membership: |
|