Assay Details
| Gene Symbol: MFRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115960.2 | XP_014971446.2 | 3026 | 63 | ||
| XM_028834071.1 | XP_028689904.1 | 3026 | 63 | |||
| XM_028834072.1 | XP_028689905.1 | 3002 | 63 | |||
| XM_028834073.1 | XP_028689906.1 | 2900 | 63 | |||
| GenBank mRNA | JV072825.1 | - | 77 | 63 | ||
More Information
Set Membership: |
|

MFRP
Genomic Map