Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836717.1 | XP_028692550.1 | 2973 | 70 | ||
| XM_028836718.1 | XP_028692551.1 | 1289 | 70 | |||
| XM_028836719.1 | XP_028692552.1 | 1211 | 70 | |||
| XM_028836720.1 | XP_028692553.1 | 1146 | 70 | |||
| XM_028836721.1 | XP_028692554.1 | 3058 | 70 | |||
| XM_028836722.1 | XP_028692555.1 | 1055 | 70 | |||
| XM_028836723.1 | XP_028692556.1 | 1204 | 70 | |||
| XM_028836724.1 | XP_028692557.1 | 1127 | 70 | |||
| XM_028836725.1 | XP_028692558.1 | 1044 | 70 | |||
| XM_028836726.1 | XP_028692559.1 | 1300 | 70 | |||
| XM_028836727.1 | XP_028692560.1 | 1223 | 70 | |||
| XM_028836728.1 | XP_028692561.1 | 1065 | 70 | |||
| XM_028836729.1 | XP_028692562.1 | 1215 | 70 | |||
| XM_028836730.1 | XP_028692563.1 | 2983 | 70 | |||
| XM_028836731.1 | XP_028692564.1 | 1049 | 70 | |||
| GenBank mRNA | JU323131.1 | - | 1589 | 70 | ||
| JU471614.1 | - | 1589 | 70 | |||
| JV044898.1 | - | 1562 | 70 | |||
| JV044899.1 | - | 1560 | 70 | |||
| JV565354.1 | - | 1594 | 70 | |||
More Information
Set Membership: |
|

AMZ2
Genomic Map