Assay Details
| Gene Symbol: DTX3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112746.4 | XP_001112746.2 | 2125 | 102 | ||
| XM_015129997.2 | XP_014985483.2 | 1947 | 102 | |||
| GenBank mRNA | JU474443.1 | - | 1972 | 102 | ||
More Information
Set Membership: |
|

DTX3L
Genomic Map