Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123614.2 | XP_014979100.1 | 382 | 85 | ||
| XM_028839046.1 | XP_028694879.1 | 523 | 85 | |||
| GenBank mRNA | JV073488.1 | - | 252 | 85 | ||
More Information
Set Membership: |
|

ZNF527
Genomic Map