Assay Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125072.2 | XP_014980558.2 | 546 | 102 | ||
| GenBank mRNA | JV496586.1 | - | 123 | 102 | ||
| JV708263.1 | - | 113 | 102 | |||
More Information
Set Membership: |
|

ZNF550
Genomic Map